Goggles, Carbonvision
Supplier: Uvex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
uvex carbonvision
111-2535EA 34.3 GBP
111-2535 111-0569 UVEX9307.276
Goggles, Carbonvision
Goggles
Modern materials and manufacturing technologies enable the design of frames which combine reduced size and a high mechanical resistance.
  • TPU layer is permanently fixed to the frame, ensuring a perfect individual fit; easy to clean
  • Lightweight enabling the wearer to work for longer periods without any distractions and problems
  • Flexible soft components adapt perfectly to the face ensuring a perfect fit
  • Convenient clip to remove headband before cleaning
These goggles are recommended for protection against dust, liquid splashes, and mechanical hazards, in combination with disposable respirators and helmets.
Uvex supravision extreme coating:
- Permanent anti-fog effect forms a thin film of water without weakening the visibility
- Extremely scratch-resistant, resistant to chemicals and easy to clean
Weight: 46 g
Certifications: EN 166, EN 170
Order Now
Learn more