Italy
Order Entry
Accessories for VWR® INCU-Line®, Large Capacity Cooled Incubator IL 500R
Accessories for VWR® INCU-Line®, Large Capacity Cooled Incubator IL 500R
Supplier:  VWR Collection
Certificates
Accessories for VWR® INCU-Line®, Large Capacity Cooled Incubator IL 500R
Supplier:  VWR Collection
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • Wire coated shelf, with slides set, 467×565 mm, max. shelf workload: 20 kg
    Wire coated shelf, with slides set, 467×565 mm, max. shelf workload: 20 kg
    390-1706
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 93,00 €
    93,00 €
    1 * 1 pezzi
       93,00 €
    Local Article Number: VWRI390-1706
    Specifications:
    Specifications
    Environmentally Preferable
  • Stainless steel perforated shelf, with slides set, 467×565 mm, max. shelf workload: 20 kg
    Stainless steel perforated shelf, with slides set, 467×565 mm, max. shelf workload: 20 kg
    390-1707
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 150,00 €
    150,00 €
    1 * 1 pezzi
       150,00 €
    Local Article Number: VWRI390-1707
    Specifications:
    Specifications
    Environmentally Preferable