Stampa…
microPHAZIR™ RX Analyser
Fornitore: Thermo Fisher Scientific
microPHAZIR™
TSPA800-01401-01EA 79210 EUR
TSPA800-01401-01
microPHAZIR™ RX Analyser
Spectroscopic Instruments Spettofotometri FTIR e IR
The Thermo Scientific microPHAZIR™ RX analyser is a handheld Near-Infrared (NIR) instrument designed for rapid on-site pharmaceutical material identification and analysis.
  • Save time and increase inspection
  • Handheld design allows operators to perform analysis right in the warehouse
  • Eliminate operator and material exposure
  • Measures directly through plastic drum liners and glass containers
  • 100% container inspection
  • Convenient, non-invasive, sampling approach permits 100% container inspection without the proportional costs
The microPHAZIR ™ RX analyser provides pharmaceutical manufacturers with a portable tool to meet increasing regulatory requirements, improve product quality, and reduce manufacturing costs. The analyser takes the power of NIR out of the laboratory and into the warehouse to cut costs, ensure product quality through increased testing frequency, while also reducing supply chain risk. The 1,25 kg analyser is battery powered and completely self-contained for truly portable analysis.
Interchangeable, rechargeable 5+ hour lithium-ion battery pack. AC battery charger included. Battery pack recharge time <2 hours. System includes two batteries. One-minute measurement. High-strength, dust-proof/splash-proof plastic housing. Tungsten light bulb light source.
Certificazioni: USP <1119> and EP 2.2.48 compliant; supports 21CFR Part 11 compliant processes.
Informazioni per l'ordine: For more information on available options and accessories, contact your Avantor representative.