Order Entry
Italy
ContactUsLinkComponent
Quanti-Cult Plus™ Enterococcus faecalis ATCC™ 29212™
Quanti-Cult Plus™ Enterococcus faecalis ATCC™ 29212™
Catalog # OXOIR4717030
CAS Number:  
undefined
Quanti-Cult Plus™ Enterococcus faecalis ATCC™ 29212™
Catalog # OXOIR4717030
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Description:
    Enterococcus faecalis ATCC™ 29212™
Specifications
About this item
Quanti-Cult Plus™ Enterococcus faecalis ATCC™ 29212™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
  • No serial dilution required
  • Delivers less-than100 colony forming units (CFUs) in a single 0,1 ml inoculum
  • 10 tests/reconstituted vial; 100 tests/kit
  • Ready for rehydration
  • For use in quality control procedures
Packaging: Includes 10 preserved organism vials and 10 rehydration vials.