Italy
Order Entry
Antistatic Waste Bags
Antistatic Waste Bags
Supplier:  DESCO
Certificates
Antistatic Waste Bags
Supplier:  DESCO
Product Information
  • Catalog #
    Supplier #
    Colour
    Capacity
    W×L
    Pk
    Availability
    Price
  • Antistatic waste bag
    Antistatic waste bag
    129-2629
    239235
    Blue
    110 L
    460×970 mm
    100
    Log in for availability
    Log in for availability
    1 * 100 pezzi
    1 * 100 pezzi - 58,30 €
    58,30 €
    1 * 100 pezzi
       58,30 €
    Supplier Article Number: 239235
    Local Article Number: DESC239235
    Specifications:
    Specifications
    Environmentally Preferable
  • Antistatic waste bag
    Antistatic waste bag
    129-2630
    239245
    Red
    110 L
    460×970 mm
    100
    Log in for availability
    Log in for availability
    1 * 100 pezzi
    1 * 100 pezzi - 59,80 €
    59,80 €
    1 * 100 pezzi
       59,80 €
    Supplier Article Number: 239245
    Local Article Number: DESC239245
    Specifications:
    Specifications
    Environmentally Preferable