Italy
Order Entry
Hand-Operated Drum Pumps, Steel
Hand-Operated Drum Pumps, Steel
undefined
Hand-Operated Drum Pumps, Steel
  • Tube ext.Ø, inlet:
  • 0,625"
  • 0,625"
  • 0,625"
  • 0,625"
  • 0,625"
  • 0,625"
  • Tube ext.Ø, outlet:
  • 0,5"
  • 0,5"
  • 0,5"
  • 0,5"
  • 1"
  • 1"
  • Tube length, inlet:
  • 23"
  • 23"
  • 37"
  • 37"
  • Wetted materials:
  • 316 SS body; PTFE packing, piston, inlet tube
  • Carbon steel body; HDPE packing, piston, inlet tube
  • 317 SS body; PTFE packing, piston, inlet tube
  • Carbon steel body; HDPE packing, piston, inlet tube
  • Steel, polyethylene
  • 304 SS, PTFE
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 3 Months
  • 3 Months
  •  
    Flow rate
    Pk
  • Hand-operated drum pump, 316 SS body, 16 strokes/gallon
    Hand-operated drum pump, 316 SS body, 16 strokes/gallon
    07079-20
    16 strokes/gal.
    1
    1 * 1 pezzi
    1 * 1 pezzi - 175,00 €
    175,00 €
    1 * 1 pezzi
       175,00 €
     : 07079-20
     : MFLB07079-20
    Accessories
    Specifications
    Environmentally Preferable
    Tube ext.Ø, inlet
    0,625"
    Tube ext.Ø, outlet
    0,5"
    Tube length, inlet
    23"
    Wetted materials
    316 SS body; PTFE packing, piston, inlet tube
    Warranty
    1 year
  • Hand-operated drum pump, steel body, 16 strokes/gallon
    Hand-operated drum pump, steel body, 16 strokes/gallon
    07079-25
    16 strokes/gal.
    1
    1 * 1 pezzi
    1 * 1 pezzi - 39,60 €
    39,60 €
    1 * 1 pezzi
       39,60 €
     : 07079-25
     : MFLB07079-25
    Accessories
    Specifications
    Environmentally Preferable
    Tube ext.Ø, inlet
    0,625"
    Tube ext.Ø, outlet
    0,5"
    Tube length, inlet
    23"
    Wetted materials
    Carbon steel body; HDPE packing, piston, inlet tube
    Warranty
    1 year
  • Hand-operated drum pump, 316 SS body, 8 strokes/gallon
    Hand-operated drum pump, 316 SS body, 8 strokes/gallon
    07079-30
    8 strokes/gal.
    1
    1 * 1 pezzi
    1 * 1 pezzi - 223,00 €
    223,00 €
    1 * 1 pezzi
       223,00 €
     : 07079-30
     : MFLB07079-30
    Accessories
    Specifications
    Environmentally Preferable
    Tube ext.Ø, inlet
    0,625"
    Tube ext.Ø, outlet
    0,5"
    Tube length, inlet
    Wetted materials
    317 SS body; PTFE packing, piston, inlet tube
    Warranty
    1 year
  • Hand-operated drum pump, steel body, 8 strokes/gallon
    Hand-operated drum pump, steel body, 8 strokes/gallon
    07079-35
    8 strokes/gal.
    1
    1 * 1 pezzi
    1 * 1 pezzi - 61,20 €
    61,20 €
    1 * 1 pezzi
       61,20 €
     : 07079-35
     : MFLB07079-35
    Accessories
    Specifications
    Environmentally Preferable
    Tube ext.Ø, inlet
    0,625"
    Tube ext.Ø, outlet
    0,5"
    Tube length, inlet
    Wetted materials
    Carbon steel body; HDPE packing, piston, inlet tube
    Warranty
    1 year
  • Hand-Operated Drum Pump, Steel/PE, 5 Strokes/Gallon; 2" IPS Bung
    Hand-Operated Drum Pump, Steel/PE, 5 Strokes/Gallon; 2" IPS Bung
    07079-80
    5 strokes/gal.
    1
    1 * 1 pezzi
    1 * 1 pezzi - 58,90 €
    58,90 €
    1 * 1 pezzi
       58,90 €
     : 07079-80
     : MFLB07079-80
    Accessories
    Specifications
    Environmentally Preferable
    Tube ext.Ø, inlet
    0,625"
    Tube ext.Ø, outlet
    1"
    Tube length, inlet
    37"
    Wetted materials
    Steel, polyethylene
    Warranty
    3 Months
  • Drum Pump, 5 Strokes/Gallon, 304 SS and PTFE
    Drum Pump, 5 Strokes/Gallon, 304 SS and PTFE
    07079-90
    5 strokes/gal.
    1
    1 * 1 pezzi
    1 * 1 pezzi - 148,00 €
    148,00 €
    1 * 1 pezzi
       148,00 €
     : 07079-90
     : MFLB07079-90
    Accessories
    Specifications
    Environmentally Preferable
    Tube ext.Ø, inlet
    0,625"
    Tube ext.Ø, outlet
    1"
    Tube length, inlet
    37"
    Wetted materials
    304 SS, PTFE
    Warranty
    3 Months