Italy
Order Entry
Male Threaded Straight Nipple, 3/4" NPT(M), PTFE
  MFLX34030-05
CE certificate
undefined
Male Threaded Straight Nipple, 3/4" NPT(M), PTFE
  MFLX34030-05
CE certificate
77,50 €
  1 * 1 pezzi
  • Connection Type:
    Threaded Union
  • Flow pattern:
    Straight
  • Thread size NPT(M) (port 1):
    ¾"
  • Thread size NPT(M) (port 2):
    ¾"
  • Description:
    Male Threaded Straight Nipple, ¾" NPT(M), PTFE
  • Sterilisation:
    Autoclave or ethylene oxide (ETO)
  • Warranty:
    2 years
  • Pk:
    1
  • Fitting material:
    PTFE
Chemically-inert PTFE handles most aggressive chemicals.
  • Constructed of durable PTFE
Chemically inert, PTFE offers outstanding resistance for any use, especially for critical high-purity applications. When subjected to higher pressure, PTFE as a material has a tendency to creep over time. As a result, these fittings work best at lower operating pressures (110 psi maximum operating pressure at ambient temperature for smaller sizes). All fittings are made from virgin PTFE material.
  • Connection Type:
    Threaded Union
  • Flow pattern:
    Straight
  • Thread size NPT(M) (port 1):
    ¾"
  • Thread size NPT(M) (port 2):
    ¾"
  • Description:
    Male Threaded Straight Nipple, ¾" NPT(M), PTFE
  • Environmentally Preferable:
  • Sterilisation:
    Autoclave or ethylene oxide (ETO)
  • Warranty:
    2 years
  • Pk:
    1
  • Fitting material:
    PTFE