Italy
Order Entry
Dual and Quad-Head Diaphragm Pumps
undefined
Dual and Quad-Head Diaphragm Pumps
  • Duty cycle continuous:
  • Yes
  • Yes
  • Inlet connection:
  • 1/4" hose barb
  • 1/4" hose barb
  • Max. temperature:
  • 80 °C (176 °F)
  • 80 °C (176 °F)
  • Motor type:
  • Brushless DC
  • Brushless DC
  • Outlet connection:
  • 1/4" hose barb
  • 1/4" hose barb
  • Power pigtail connection:
  • Yes
  • Yes
  • Run dry:
  • Yes
  • Yes
  • Self-priming:
  • Yes
  • Yes
  • Wetted materials:
  • Polypropylene, Silicone
  • Polypropylene, Silicone
  • Warranty:
  • 1 year
  • 1 year
  •  
    Maximum Flow Rate
    Power
    Height
    Length
    Width
    Pk
  • Quad-Head Diaphragm Pump, for Gases, 29,9 L/min; 12 VDC
    Quad-Head Diaphragm Pump, for Gases, 29,9 L/min; 12 VDC
    78182-30
    29,9 L/min (7,9 gal./min)
    12 VDC
    44,5 mm
    14,61 cm
    10,8 cm
    1
    1 * 1 pezzi
    1 * 1 pezzi - 422,00 €
    422,00 €
    1 * 1 pezzi
       422,00 €
     : 78182-30
     : MFLB78182-30
    Specifications
    Environmentally Preferable
    Duty cycle continuous
    Yes
    Inlet connection
    1/4" hose barb
    Max. temperature
    80 °C (176 °F)
    Motor type
    Brushless DC
    Outlet connection
    1/4" hose barb
    Power pigtail connection
    Yes
    Run dry
    Yes
    Self-priming
    Yes
    Wetted materials
    Polypropylene, Silicone
    Warranty
    1 year
  • Quad-Head Diaphragm Pump, for Fluids, 4,9/3,2 L/min; 12 VDC
    Quad-Head Diaphragm Pump, for Fluids, 4,9/3,2 L/min; 12 VDC
    78182-15
    4,9 L/min (1,3 gal./min)
    12 VDC
    44,5 mm
    14,61 cm
    10,8 cm
    1
    1 * 1 pezzi
    1 * 1 pezzi - 418,00 €
    418,00 €
    1 * 1 pezzi
       418,00 €
     : 78182-15
     : MFLB78182-15
    Specifications
    Environmentally Preferable
    Duty cycle continuous
    Yes
    Inlet connection
    1/4" hose barb
    Max. temperature
    80 °C (176 °F)
    Motor type
    Brushless DC
    Outlet connection
    1/4" hose barb
    Power pigtail connection
    Yes
    Run dry
    Yes
    Self-priming
    Yes
    Wetted materials
    Polypropylene, Silicone
    Warranty
    1 year