Stampa…
384-well Tips, 70 µl, SLAS rack, Axygen®
Fornitore: Corning
AXYGVT-384-70-LRSEA 6520 EUR
AXYGVT-384-70-LRS
384-well Tips, 70 µl, SLAS rack, Axygen®
Puntali per pipette
Axygen® offers a comprehensive line of tips for Agilent®/Velocity11 automation systems.
  • Maximum Recovery technology for ultra-low retention
  • RNase-/DNase-free and nonpyrogenic
  • Clear polypropylene tips
  • Non filtered
  • Sterile
  • Low Retention
These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
Compatibility: Agilent®/Velocity VPrep® and Bravo 384ST, 96ST and 16ST head.