Italy
Order Entry
VWR® Real Time Electrophoresis Systems
VWR® Real Time Electrophoresis Systems
Supplier:  VWR Collection
Certificates
undefined
VWR® Real Time Electrophoresis Systems
Supplier:  VWR Collection
Specifications
  • Catalog No:
  • 700-1472
  • 700-1473
  • Buffer volume:
  • 200 - 225 ml
  • 200 - 225 ml
  • Tank W×D×H:
  • 120×110×45 mm (inner dimensions)
  • 120×110×45 mm (inner dimensions)
Specifications
230 V Unit without power supply
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • VWR® Real Time electrophoresis gel tank
    VWR® Real Time electrophoresis gel tank
    700-1472
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 939,00 €
    939,00 €
    1 * 1 pezzi
       939,00 €
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Buffer volume
    200 - 225 ml
    Tank W×D×H
    120×110×45 mm (inner dimensions)
230 V Unit with power supply
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • VWR® Real Time electrophoresis gel system
    VWR® Real Time electrophoresis gel system
    700-1473
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 1 150,00 €
    1 150,00 €
    1 * 1 pezzi
       1 150,00 €
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Buffer volume
    200 - 225 ml
    Tank W×D×H
    120×110×45 mm (inner dimensions)