Stampa…
Hai cercato: Sistemi di associazione e marcatura delle proteine
160  risultati sono stati trovati
SearchResultCount:"160"
Sort Results
Visualizzazione a lista Nuova Visualizzazione
La tua valutazione
Codice catalogo: (M336-1L)
Fornitore: VWR Chemicals
Codice articolo locale: VWRCM336-1L
Descrizione: Antibody Signal Enhancer is a Western blotting antibody diluent that increases intensity of protein detection up to 5-fold compared to conventional bl...
UOM: 1 * 1 L
Codice catalogo: (10103.)
Fornitore: Biotium
Codice articolo fornitore: 10103
Codice articolo locale: BTIU10103
Descrizione: Luciferin is a substrate for the enzyme firefly luciferase.
UOM: 1 * 5 mg
Codice catalogo: (APOSBIL3118-100ML)
Fornitore: Apollo Scientific
Codice articolo fornitore: BIL3118-100ML
Codice articolo locale: APOSBIL3118-100ML
Descrizione: Liquid product colourless to slightly yellow or greenish odourless pH 7,3 to 7,5 boiling point 104 °C. Stabiliser for controls and protein standards.
UOM: 1 * 100 mL
Fornitore: Biotium
Descrizione: Label your antibody with one of our superior CF® Dyes in 30 minutes without a purification step. Labelling tolerates many common buffer components inc...
Fornitore: Biotium
Descrizione: Annexin V fluorescent conjugates can be used to detect apoptotic cells. Annexin V is available conjugated to a wide variety of dyes.
Fornitore: Biotium
Descrizione: Phalloidin is a toxin isolated from the deadly Amanita phalloides mushroom. It is a bicyclic peptide that binds specifically to F-actin. Fluore...
Fornitore: Biotium
Descrizione: Kits for increasing immunofluorescence sensitivity using HRP-catalysed tyramide signal amplification with the choice of next-generation fluorescent CF...
Fornitore: Biotium
Descrizione: Mix-n-Stain™ FITC antibody labelling kits utilise a technology that allows rapid antibody labelling with green fluorescent dye (FITC) without a purifi...
Fornitore: Biotium
Descrizione: Biotium offers reactive biotin compounds with a variety of reactive functional groups.
Fornitore: Biotium
Descrizione: A wheat germ agglutinin (WGA) lectin labelled with Biotium’s CF® Dyes or HRP. It is a carbohydrate-binding lectin that has high affinity for sialic ac...
Fornitore: Biotium
Descrizione: Cholera toxin subunit B conjugated to Biotium's next-generation fluorescent CF™ dyes.
Fornitore: Biotium
Descrizione: ExoBrite™ CTB EV Staining Kits were designed to overcome some of the challenges of EV detection, particularly in flow cytometry. ExoBrite™ CTB EV Stai...
Fornitore: Biotium
Descrizione: The simplicity and convenience of Mix-n-Stain™ labelling of nanobodies with fluorescent CF® dyes or biotin.
Fornitore: Biotium
Descrizione: Reactive dyes containing an amine functional group.
Fornitore: Biotium
Descrizione: Reactive dyes for labeling cysteine residues in proteins and other thiol molecules. For monitoring and validating the sterilisation process.
Fornitore: Biotium
Descrizione: Easily label SNAP-tag®, CLIP-tag™, HaloTag®, or other ligands with fluorescent dyes, no purification needed. Cell membrane-permeant and impermeant dye...
Prezzo a richiesta
La disponibilità per questo articolo è limitata, ma potrebbe essere disponibile in un magazzino vicino a voi. Si prega di assicurarsi che si è effettuato l'accesso al sito, in modo che ledisponibilità possano essere visualizzati. Se il call è ancora visualizzato e hai bisogno di assistenza, si prega di telefonare a 1-800-932 - 5000.
La disponibilità per questo articolo è limitata, ma potrebbe essere disponibile in un magazzino vicino a voi. Si prega di assicurarsi che si è effettuato l'accesso al sito, in modo che ledisponibilità possano essere visualizzati. Se il call è ancora visualizzato e hai bisogno di assistenza, si prega di telefonare a 1-800-932 - 5000.
Puoi ordinare questo prodotto. Se necessario, ti contatteremo per richiedere licenza, autorizzazione o dichiarazione d'uso per procedere con la fornitura.
Puoi ordinare questo prodotto. Se necessario, ti contatteremo per richiedere licenza, autorizzazione o dichiarazione d'uso per procedere con la fornitura.
Questo prodotto è stato bloccato dalla vostra organizzazione. Si prega di contattare il vostro responsabile acquisti per ulteriori informazioni.
Il prodotto originale non è più disponibile. L'articolo suggerito è disponibile
Il prodotto marcato con questo simbolo non è più disponibile o vendibile fino ad esaurimento scorte. Alternative possono essere disponibili utilizzando il numero di catalogo VWR. Per maggiori informazioni può contattare il nostro customer service al 023320311 o inviando una email all'indirizzo ebusiness.it@vwr.com
1 - 16 of 160
no targeter for Bottom