Stampa…
Capillary viscometer, iVisc
Fornitore: Lauda
iVisc
616-0098EA 0 EUR
616-0098
Capillary viscometer, iVisc
Viscosimetri Viscometri Capillari
The iVisc is a compact instrument, for operation with a PC computer, that is designed for use with a wide range of standard capillaries (Ubbelohde, Cannon-Fenske and micro-Otswald).
  • Simple to operate with intuitive software and 'plug and play installation'
  • Self adaptive, intelligent NIR meniscus sensing even for problematic liquids
  • Only one cable (USB) for control and power supply
  • Connection of up to two iVisc per computer
Insert the filled capillary and the software co-ordinates all the stage required for measurement and evaluation. With a constant temperature bath and glass capillary, the iVisc has applications across a broad range of industries from pharmaceutical R&D, food QA and petrochemicals to polymer producers and plastic processors.
Certificazioni: ISO 3104 and 2909; ISO 1628 and 307; DIN 51562 and 53728; ASTM