Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Italy
Order Entry
Order Status
Your Account
Cart
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0,00 €
Order Entry
Home
3-Fluoro-2-butanone
3-Fluoro-2-butanone 97%
3-Fluoro-2-butanone 97%
Flammable
3-Fluoro-2-butanone 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Flammable
3-Fluoro-2-butanone 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
814-79-9
:
Formula:
C₄H₇FO
MDL Number:
MFCD01862088
:
90.1 g/mol
Boiling Pt:
74…76 °C
Storage Temperature:
Ambient
UN:
1224
ADR:
3,II
:
GHS02
Product Information
Catalog #
Supplier #
Pack type
Pk
Availability
Price
APOSPC9588-1G
PC9588-1G
Bottle
1 g
Log in
for availability
Log in
for availability
1 * 1 g
1 * 1 g - 198,00 €
198,00 €
-
+
Add to cart
1 * 1 g
198,00 €
-
+
Add to cart
APOSPC9588-5G
PC9588-5G
Bottle
5 g
Log in
for availability
Log in
for availability
1 * 5 g
1 * 5 g - 613,00 €
613,00 €
-
+
Add to cart
1 * 5 g
613,00 €
-
+
Add to cart