1,2-Dichloro-3,3,3-trifluoro-2-(trifluoromethyl)propane 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:138690-25-2
:
Formula:C₄H₂Cl₂F₆
MDL Number:MFCD01862044
:234.96 g/mol
Boiling Pt:78…79 °C
:GHS07
Storage Temperature:Ambient
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: PC8346-1GLocal Article Number: APOSPC8346-1G
- Supplier Article Number: PC8346-5GLocal Article Number: APOSPC8346-5G
- Supplier Article Number: PC8346-25GLocal Article Number: APOSPC8346-25G