1-(2,5-Difluorophenyl)biguanide hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:66088-53-7
:
Formula:C₈H₁₀ClF₂N₅
MDL Number:MFCD03094448
Melting Pt:225…230 °C
:GHS07
Storage Temperature:Ambient
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: PC7815-5GLocal Article Number: APOSPC7815-5G
- Supplier Article Number: PC7815-25GLocal Article Number: APOSPC7815-25G