4,5-Difluoro-2-nitroaniline 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:78056-39-0
:
Formula:C₆H₄F₂N₂O₂
MDL Number:MFCD00010876
:174.11 g/mol
Melting Pt:107…108 °C
:GHS07
Storage Temperature:Ambient
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: PC6082-5GLocal Article Number: APOSPC6082-5G
- Supplier Article Number: PC6082-100GLocal Article Number: APOSPC6082-100G