3-[4-Fluoro-3-(trifluoromethyl)phenyl]propionic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
:
Formula:C₁₀H₈F₄O₂
MDL Number:MFCD09025359
:236.16 g/mol
Storage Temperature:Ambient
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: PC302280-1GLocal Article Number: APOSPC302280-1G
- Supplier Article Number: PC302280-5GLocal Article Number: APOSPC302280-5G
- Supplier Article Number: PC302280-10GLocal Article Number: APOSPC302280-10G