Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Italy
Order Entry
Order Status
Your Account
Cart
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0,00 €
Order Entry
Home
2-Bromo-6-cyanopyrazine
2-Bromo-6-cyanopyrazine 97%
2-Bromo-6-cyanopyrazine 97%
2-Bromo-6-cyanopyrazine 97%
Supplier:
Apollo Scientific
Certificates
Expand image
2-Bromo-6-cyanopyrazine 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
859064-02-1
:
Formula:
C₅H₂BrN₃
MDL Number:
MFCD11109800
:
183.9900 g/mol
Storage Temperature:
Ambient
Product Information
Catalog #
Supplier #
Pack type
Pk
Availability
Price
APOSOR904498-1G
OR904498-1G
Bottle
1 g
Log in
for availability
Log in
for availability
1 * 1 g
1 * 1 g - 104,00 €
104,00 €
-
+
Add to cart
1 * 1 g
104,00 €
-
+
Add to cart
APOSOR904498-5G
OR904498-5G
Bottle
5 g
Log in
for availability
Log in
for availability
1 * 5 g
1 * 5 g - 404,00 €
404,00 €
-
+
Add to cart
1 * 5 g
404,00 €
-
+
Add to cart