Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Italy
Order Entry
Order Status
Your Account
Cart
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0,00 €
Order Entry
Home
4-(1-Hydroxyethyl)biphenyl
4-(1-Hydroxyethyl)biphenyl 98%
4-(1-Hydroxyethyl)biphenyl 98%
Irritant
4-(1-Hydroxyethyl)biphenyl 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-(1-Hydroxyethyl)biphenyl 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
3562-73-0
:
Formula:
C₁₄H₁₄O
MDL Number:
MFCD00016859
:
198.27 g/mol
Melting Pt:
95…97 °C
:
GHS07
Storage Temperature:
Ambient
Product Information
Catalog #
Supplier #
Pack type
Pk
Availability
Price
APOSOR451303-5G
OR451303-5G
Bottle
5 g
Log in
for availability
Log in
for availability
1 * 5 g
1 * 5 g - 30,60 €
30,60 €
-
+
Add to cart
1 * 5 g
30,60 €
-
+
Add to cart
APOSOR451303-25G
OR451303-25G
Bottle
25 g
Log in
for availability
Log in
for availability
1 * 25 g
1 * 25 g - 104,00 €
104,00 €
-
+
Add to cart
1 * 25 g
104,00 €
-
+
Add to cart
APOSOR451303-100G
OR451303-100G
Bottle
100 g
Log in
for availability
Log in
for availability
1 * 100 g
1 * 100 g - 347,00 €
347,00 €
-
+
Add to cart
1 * 100 g
347,00 €
-
+
Add to cart