Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Italy
Order Entry
Order Status
Your Account
Cart
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0,00 €
Order Entry
Home
1-(3-Methoxyphenyl)piperidine
1-(3-Methoxyphenyl)piperidine 98%
1-(3-Methoxyphenyl)piperidine 98%
1-(3-Methoxyphenyl)piperidine 98%
Supplier:
Apollo Scientific
Certificates
Expand image
1-(3-Methoxyphenyl)piperidine 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
32040-06-5
:
Formula:
C₁₂H₁₇NO
MDL Number:
MFCD00094104
:
191.269 g/mol
Storage Temperature:
Ambient
Product Information
Catalog #
Supplier #
Pack type
Pk
Availability
Price
APOSOR900221-1G
OR900221-1G
Bottle
1 g
Log in
for availability
Log in
for availability
1 * 1 g
1 * 1 g - 145,00 €
145,00 €
-
+
Add to cart
1 * 1 g
145,00 €
-
+
Add to cart
APOSOR900221-5G
OR900221-5G
Bottle
5 g
Log in
for availability
Log in
for availability
1 * 5 g
1 * 5 g - 328,00 €
328,00 €
-
+
Add to cart
1 * 5 g
328,00 €
-
+
Add to cart