Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Italy
Order Entry
Order Status
Your Account
Cart
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0,00 €
Order Entry
Home
3-Bromo-4-iodo-1H-pyrazole
3-Bromo-4-iodo-1H-pyrazole
3-Bromo-4-iodo-1H-pyrazole
Irritant
3-Bromo-4-iodo-1H-pyrazole
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3-Bromo-4-iodo-1H-pyrazole
Supplier:
Apollo Scientific
About this item
CAS Number:
27258-15-7
:
Formula:
C₃H₂BrIN₂
MDL Number:
MFCD27988092
:
272.87 g/mol
:
GHS07
Storage Temperature:
Ambient
Product Information
Catalog #
Supplier #
Pack type
Pk
Availability
Price
APOSOR307261-1G
OR307261-1G
Bottle
1 g
Log in
for availability
Log in
for availability
1 * 1 g
1 * 1 g - 102,00 €
102,00 €
-
+
Add to cart
1 * 1 g
102,00 €
-
+
Add to cart
APOSOR307261-5G
OR307261-5G
Bottle
5 g
Log in
for availability
Log in
for availability
1 * 5 g
1 * 5 g - 203,00 €
203,00 €
-
+
Add to cart
1 * 5 g
203,00 €
-
+
Add to cart