(+/-)-cis-2-Methyl-1,2,3,4-tetrahydro-quinoline-4-carboxylic acid methyl ester
Supplier: Apollo Scientific
Certificates
About this item
:
Formula:C₁₂H₁₅NO₂
MDL Number:MFCD24447594
:205.253 g/mol
Storage Temperature:Ambient
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: OR303282-500MGLocal Article Number: APOSOR303282-500MG
- Supplier Article Number: OR303282-1GLocal Article Number: APOSOR303282-1G