Stampa…
LEUCOGNOST® Fixing mixture, Sigma-Aldrich®
Fornitore: Merck
LEUCOGNOST®
1.12327.0000EA 0 EUR
1.12327.0000
LEUCOGNOST® Fixing mixture, Sigma-Aldrich®
Coloranti Reagenti per microscopia e colorazione
Fixing reagent for enzyme cytochemistry
LEUCOGNOST® fixing mixture is especially developed for the fixation of blood and bone marrow smears using the various LEUCOGNOST® kits. LEUCOGNOST® fixing mixture optimally protects enzyme activities, and the reaction times of the different working solutions are specially matched to the fixing mixture. It is developed and registered as CE / IVD product for applications in diagnostics.
Can be used with the following LEUCOGNOST® kits
Art. Nr. 116300 LEUCOGNOST® ALPA 1 Kit
Art. Nr. 116301 LEUCOGNOST® EST 1 Kit
Art. Nr. 116302 LEUCOGNOST® PAS 1 Kit
Art. Nr. 116303 LEUCOGNOST® POX 1 Kit
Art. Nr. 116304 LEUCOGNOST® SP 1 Kit
Art. Nr. 116198 LEUCOGNOST® NASDCL 1 Kit