
Oxaliplatin
TCIAO0372-100MG
: TCI
:
- Pk:100 mg
- Seq:2440
- Appearance:White to Almost white powder to crystal
- Elemental analysis (Nitrogen):6.70 to 7.40 %
:
:397.29 g/mol
:14,06912
:GHS08
:Refrigerator
:61825-94-3
:C₈H₁₄N₂O₄Pt
:MFCD00866327