Cerium tetrafluoride hydrate
Fornitore: Thermo Scientific
|
Pericolo
|
38580.09EA
76.6
EUR
38580.09
Cerium tetrafluoride hydrate
Cerium tetrafluoride hydrate
Formula:
CeF₄·H₂O Peso molecolare: 234,12 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00049422 CAS Number: 60627-09-0 UN: 3288 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...