Stampa…
Acqua, SupraSolv® for gas chromatography, Supelco®
Fornitore: Merck
M-Clarity™ quality level = MQ100
1.02699.1000EA 54 EUR
1.02699.1000 1.02699.2500
Acqua, SupraSolv® for gas chromatography, Supelco®
Acqua
Formula: H₂O
Peso molecolare: 18,02 g/mol
Punto di ebollizione: 100 °C (1013 hPa)
Punto di fusione: 0 °C
Densità: 1 g/cm³ (4 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 12,10175
For your Gaz or HPLC chromatography method
ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
Visualizza i codici
For your Gaz or HPLC chromatography method
ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation