Italy
Order Entry
VWR®, Certified Reference Material, Cleveland Open Cup flash point
VWR®, Certified Reference Material, Cleveland Open Cup flash point
Supplier:  VWR Chemicals
Certificates
undefined
VWR®, Certified Reference Material, Cleveland Open Cup flash point
Supplier:  VWR Chemicals
Certificates
Related Information
Product Information
  • Catalog #
    Flash point
    Pack type
    Pk Info
    Pk
    Availability
    Price
  • Certified Reference Material, Cleveland Open Cup flash point
    Certified Reference Material, Cleveland Open Cup flash point
    111,0 °C
    Glass bottle
    3×80 ml
    1 SET
    Log in for availability
    1 * 1 SET
    1 * 1 SET - 403,00 €
    403,00 €
    1 * 1 SET
       403,00 €
  • Certified Reference Material, Cleveland Open Cup flash point
    Certified Reference Material, Cleveland Open Cup flash point
    161,4 °C
    Glass bottle
    3×80 ml
    1 SET
    Log in for availability
    1 * 1 SET
    1 * 1 SET - 403,00 €
    403,00 €
    1 * 1 SET
       403,00 €
  • Certified Reference Material, Cleveland Open Cup flash point
    Certified Reference Material, Cleveland Open Cup flash point
    262,0 °C
    Glass bottle
    3×80 ml
    1 SET
    Log in for availability
    1 * 1 SET
    1 * 1 SET - 403,00 €
    403,00 €
    1 * 1 SET
       403,00 €
  • Certified Reference Material, Cleveland Open Cup flash point (lubricant)
    Certified Reference Material, Cleveland Open Cup flash point (lubricant)
    256,8 °C
    Glass bottle
    1×250 ml
    250 mL
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 292,00 €
    292,00 €
    1 * 250 mL
       292,00 €