Stampa…
Battery, Versaflo™ TR-830
Fornitore: 3M
Versaflo™
111-2548EA 1470 EUR
111-2548
Battery, Versaflo™ TR-830
Batterie
The fast-charging, intrinsically safe Versaflo™ lithium-ion battery TR 830 is intended for use with the 3M™ Versaflo™ TR 800 series PAPR system.
The battery reaches 90% charge in approximately three and a half hours and features a charge level indicator for easy visual reference. It is engineered for use in potentially explosive athmospheres and features yellow bumpers to help absorb impact.
When equipped with a new intrinsically safe battery (TR-830) and new filter, the TR-800 PAPR assembly has a run time of 6 to 10 hours depending on system configuration used. PAPR runtime will vary based on battery age, filter loading, filter or cartridge selection, and user selected airflow setting. The battery also can provide approximately 750 full charge/discharge cycle equivalents while maintaining 80% of its original capacity, which is 2X times the cycle life of standard non-IS Versaflo™ batteries TR-630 and TR-632.
Certificazioni: ATEX certified, NIOSH approved.
Informazioni per l' accessorio: Accessory TR-838 for the simple and safe attachment of the battery type TR 830 to the blower unit TR 802E. The battery attachment tool is ATEX certified and NIOSH approved. The TR-838 battery attachment tool (included) fastens the battery mechanically to the blower unit to help prevent accidental removal and reduce spark risk during use per EN 60079-11.