Stampa…
Digital refrigerator thermometer, TMX
Fornitore: Ebro
ebro®
EBRO1340-2550EA 38.5 EUR
EBRO1340-2550
Digital refrigerator thermometer, TMX
Termometri Termometri digitali
Min./max. thermometer with external probe.
  • Monitoring of two temperature zones
  • Easy to use
  • Reliable control
  • Acoustic alarm
  • Optical alarm signal
To monitor the sample temperature in the laboratory, but also in microbiological research facilities, a thermometer with minimum and maximum value display is required. In addition, all refrigerators, pharmaceuticals and vaccines, chemicals, greenhouses, blood banks, food and beverage must be monitored and storage facilities monitored. Minimum and maximum values must be recorded daily and documented manually.
To simplify the process and for easy monitoring in daily use in the field of application, the thermometer simultaneously displays the current measured value and Min / Max. The employee has all the information at a glance and can intervene directly if necessary. If a limit is exceeded, a warning tone sounds in addition to the measured value display, which alerts you to a current problem in maintaining the cold chain.
Informazioni per l'ordine: Supplied with factory calibration certificate (–20 °C, 0 °C and +60 °C).