Stampa…
HPLC <621> PT, proficiency testing material, Supelco®
Fornitore: Merck
PEP015-2X1GEA 275 EUR
PEP015-2X1G
HPLC <621> PT, proficiency testing material, Supelco®
Standard Standard per le prove valutative
This proficiency test can be used to comply with ISO 17025 requirements for proficiency testing, demonstration of capability, and analysts training for HPLC technologies by methodology described by USP <621>.
Certificazioni: This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. 