Stampa…
Organic reference standard, 3-Methylmalic acid, ≥95,0% (HPLC), Supelco®
Fornitore: Merck
69369-50MGEA 581 EUR
69369-50MG 69369-10MG
Organic reference standard, 3-Methylmalic acid, ≥95,0% (HPLC), Supelco®
Standard Standard organici
Metabolite of C5-branched dibasic acid metabolism, valine, leucine and isoleucine biosynthesis and 2-oxocarboxylic acid metabolism.