Stampa…
Pesticide reference standard, Phoxim, PESTANAL®, analytical standard, Supelco®
Fornitore: Merck
SUPL36197-100MGEA 41 EUR
SUPL36197-100MG
Pesticide reference standard, Phoxim, PESTANAL®, analytical standard, Supelco®
Phoxim may be used as an analytical reference standard for the determination of phoxim in:
  • Honeys, honeybees and pollens and food samples by QuEChERS extraction followed by analysis using liquid chromatography-electrospray ionisation-tandem mass spectrometry (LC-ESI-LC-MS/MS) combined with multiple reaction monitoring (MRM) detection
  • Egg samples by acetonitrile-based extraction and high performance liquid chromatography-diode array detection (HPLC-DAD)
  • Environmental samples by temperature-controlled ionic liquid dispersive liquid-phase extraction and HPLC combined with ultraviolet (UV) detection
It is classified under the organophosphorus group of pesticides.