Stampa…
Pesticide reference standard, Fenthion-sulfone, PESTANAL®, analytical standard, Supelco®
Fornitore: Merck
PESTANAL®
46023-10MGEA 95.9 EUR
46023-10MG
Pesticide reference standard, Fenthion-sulfone, PESTANAL®, analytical standard, Supelco®
Standard Standard organici
The metabolite of fenthion which is classified under the organophosphorus group of pesticides.
Used as an analytical reference standard for the determination of fenthion-sulfone in fruits and vegetables and other food matrices by QuEChERS extraction and low-pressure gas chromatography-time-of-flight mass spectrometry (LP-GC-TOFMS), water samples by solid-phase extraction (SPE), liquid chromatography-triple quadrupole tandem mass spectrometry (LC-QqQ-MS/MS) and LC coupled to hybrid QTOF-MS, tea samples by QuEChERS extraction, liquid-liquid extraction (LLE) cleanup and analysis using GC-MS/MS equipped with multiple reaction monitoring (MRM) detection.