Stampa…
Hotplates REC series
VELPF20700081EA 304 EUR
VELPF20700081
Hotplates REC series
The REC is a digital hot plate with white ceramic heating plate, that ensures excellent resistance to chemicals and scratches and is extremely easy to clean.
  • Innovative technopolymer body, ideal for premium resistance to acids, bases and solvents
  • Microprocessor technology ensures fast heating and provides precise temperature setting
Designed to last and equipped to ensure maximum protection against leakages with the elevated front panel, dedicated run-off grove and 'Hot Plate Warning'.