Italy
Order Entry
VISOCOLOR® Colorimetric Tests, DEHA
Supplier:  MACHEREY-NAGEL
Certificates
undefined
VISOCOLOR® Colorimetric Tests, DEHA
Supplier:  MACHEREY-NAGEL
Certificates
Specifications
  • Model:
    MANA931224
  • Platform:
    Colorimetric test kits
  • Storage temperature:
    15...25 °C
  • Shelf life (months):
    12
  • Weight:
    131,1 g (Including packaging)
Specifications
Test kits
Product Information
  • Catalog #
    Supplier #
    Contents
    Range
    Chemical method
    For unit
    Dimensions
    Availability
    Price
  • VISOCOLOR® ECO DEHA; Carbohydrazide (carbo), hydroquinone (hydro), isoascorbic acid (ISA) and methylethyl ketone oxime (MEKO) are only photometrically evaluable
    VISOCOLOR® ECO DEHA; Carbohydrazide (carbo), hydroquinone (hydro), isoascorbic acid (ISA) and methylethyl ketone oxime (MEKO) are only photometrically evaluable
    MANA931024
    931024
    Reagents for 125 tests, colour chart and accessories in a box
    0 • 0,01 • 0,03 • 0,05 • 0,10 • 0,15 • 0,20 • 0,25 • 0,30 mg/L (DEHA); 0,005 - 0,5 mg/L (DEHA); 0,01 - 0,60 mg/L (Carbo); 0,01 - 0,60 mg/L (Carbo); 0,01 - 0,60 mg/L (Carbo); 0,01 - 1,00 mg/L (Hydro); 0,01 - 1,00 mg/L (Hydro); 0,025 - 1,500 mg/L (ISA)
    Redox reaction
    PF-12Plus; Advance; Visual
    170×125×48 mm (Including packaging)
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 105,00 €
    105,00 €
    1 * 1 KIT
       105,00 €
    Supplier Article Number: 931024
    Local Article Number: MANA931024
Refill packs
Product Information
  • Catalog #
    Supplier #
    Contents
    Range
    Chemical method
    For unit
    Dimensions
    Availability
    Price
  • VISOCOLOR® ECO DEHA, refill pack
    VISOCOLOR® ECO DEHA, refill pack
    MANA931224
    931224
    Reagents for 125 tests in a box
    0 • 0,01 • 0,03 • 0,05 • 0,10 • 0,15 • 0,20 • 0,25 • 0,30 mg/L (DEHA); 0,005 - 0,5 mg/L (DEHA); 0,01 - 0,60 mg/L (Carbo)
    Redox reaction
    PF-12Plus; Advance; Visual
    170×115×70 mm (Including packaging)
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 63,90 €
    63,90 €
    1 * 1 KIT
       63,90 €
    Supplier Article Number: 931224
    Local Article Number: MANA931224