Stampa…
Chemical resistant gloves, Tychem® NT470
Fornitore: DuPont
Tychem®
112-5490EA 0 EUR
112-5490 112-5491 112-5486 112-5487 112-5488 112-5489
Chemical resistant gloves, Tychem® NT470
Guanti
These light polyvalent gloves are made of 100% nitrile and are designed to last and reduce fatigue.
  • Chemical resistance against a broad range of solvents, oils, animal fats and other chemicals
  • Mechanical resistance
  • Bisque finish
  • Ergonomic design to maximize comfort
Ideal for working safely in damp or greasy environments.
Certificazioni: EN 388:2016 (2001X), EN ISO 374-1:2016 Type A (JKLOPT), EN ISO 374-5:2016 (VIRUS); Cat. III
Food contact approved.