Industrial gloves, oil repellent, HyFlex® 11-939
Supplier: Ansell
Certificates
About this item
RIPEL technology keeps oil from making contact with the skin, keeping hands safe and dry.
- GRIP Technology delivers an average of 13% better grip performance for safer handling of wet or oily materials
- Protects against lacerations, oil and industrial fluid exposure
- Ultra-lightweight design provides dexterity and a high level of comfort
- Reinforced thumb crotch for increased protection and high durability FORTIX coating up to 12x for extended product use life
- Design and ZONZ Comfort Fit technology delivering all day comfort and silicone free
- Improved abrasion resistance up to 40% and lightest weight up to 20%
Suitable applications include: Machinery and equipment, metal fabrication, automotive, assembly and inspection of components, picking, fastening components, checking parts, adjusting panels parts components.
Certifications: EN 388:2016 (4X42B), Cat. II.
Specifications
- Catalog No:
- ANSE11-939/11
- ANSE11-939/6
- ANSE11-939/7
- ANSE11-939/8
- ANSE11-939/10
- ANSE11-939/9
- Model:
- 11-939
- 11-939
- 11-939
- 11-939
- 11-939
- 11-939
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Dyneema® Diamond Technology, nylon, Spandex and carbon fibers
- Dyneema® Diamond Technology, nylon, Spandex and carbon fibers
- Dyneema® Diamond Technology, nylon, Spandex and carbon fibers
- Dyneema® Diamond Technology, nylon, Spandex and carbon fibers
- Dyneema® Diamond Technology, nylon, Spandex and carbon fibers
- Dyneema® Diamond Technology, nylon, Spandex and carbon fibers
- Finish:
- Fully coated
- Fully coated
- Fully coated
- Fully coated
- Fully coated
- Fully coated
- Gauge:
- 18
- 18
- 18
- 18
- 18
- 18
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
- Anti-static:
- +
- +
- +
- +
- +
- +
Specifications
Product Information
- Catalog #Supplier #Glove sizeColourLengthPackedPkAvailabilityPrice
- ANSE11-939/611-939/66Black/grey212 - 261 mm12 pairs/polybag12 PAIRLog in for availability1 * 12 PAIO228,00 €Supplier Article Number: 11-939/6Local Article Number: ANSE11-939/6Specifications:
More
Specifications
Model11-939Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialDyneema® Diamond Technology, nylon, Spandex and carbon fibersFinishFully coatedGauge18Cuff styleKnit wristEN normEN 388:2016 (4X42B)Anti-static+ - ANSE11-939/711-939/77Black/grey212 - 261 mm12 pairs/polybag12 PAIRLog in for availability1 * 12 PAIO228,00 €Supplier Article Number: 11-939/7Local Article Number: ANSE11-939/7Specifications:
More
Specifications
Model11-939Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialDyneema® Diamond Technology, nylon, Spandex and carbon fibersFinishFully coatedGauge18Cuff styleKnit wristEN normEN 388:2016 (4X42B)Anti-static+ - ANSE11-939/811-939/88Black/grey212 - 261 mm12 pairs/polybag12 PAIRLog in for availability1 * 12 PAIO228,00 €Supplier Article Number: 11-939/8Local Article Number: ANSE11-939/8Specifications:
More
Specifications
Model11-939Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialDyneema® Diamond Technology, nylon, Spandex and carbon fibersFinishFully coatedGauge18Cuff styleKnit wristEN normEN 388:2016 (4X42B)Anti-static+ - ANSE11-939/911-939/99Black/grey212 - 261 mm12 pairs/polybag12 PAIRLog in for availability1 * 12 PAIO228,00 €Supplier Article Number: 11-939/9Local Article Number: ANSE11-939/9Specifications:
More
Specifications
Model11-939Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialDyneema® Diamond Technology, nylon, Spandex and carbon fibersFinishFully coatedGauge18Cuff styleKnit wristEN normEN 388:2016 (4X42B)Anti-static+ - ANSE11-939/1011-939/1010Black/grey212 - 261 mm12 pairs/polybag12 PAIRLog in for availability1 * 12 PAIO228,00 €Supplier Article Number: 11-939/10Local Article Number: ANSE11-939/10Specifications:
More
Specifications
Model11-939Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialDyneema® Diamond Technology, nylon, Spandex and carbon fibersFinishFully coatedGauge18Cuff styleKnit wristEN normEN 388:2016 (4X42B)Anti-static+ - ANSE11-939/1111-939/1111Black/grey212 - 261 mm12 pairs/polybag12 PAIRLog in for availability1 * 12 PAIO228,00 €Supplier Article Number: 11-939/11Local Article Number: ANSE11-939/11Specifications:
More
Specifications
Model11-939Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLiner materialDyneema® Diamond Technology, nylon, Spandex and carbon fibersFinishFully coatedGauge18Cuff styleKnit wristEN normEN 388:2016 (4X42B)Anti-static+