Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Italy
Order Entry
Order Status
Your Account
Cart
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0,00 €
Order Entry
Pharmaceutical secondary standard, sodium nitroprusside, (CRM), Supelco®
PHR1423-1G
:
Merck
:
Pharmaceutical secondary standard, sodium nitroprusside, (CRM), Supelco®
PHR1423-1G
:
Merck
:
108,00 €
1 * 1 g
-
+
Pk:
1 g
CAS No.:
13755-38-9
Description:
Sodium Nitroprusside
Description:
Sodium Nitroprusside