Stampa…
Safety knife, SECUNORM PROFI25
Fornitore: Martor
233-0210EA 161 EUR
MARO120.000
Safety knife, SECUNORM PROFI25
The automatic blade retraction of the SECUNORM PROFI25 guarantees a high degree of safety from cut injuries. The plastic insert of the SECUNORM PROFI25 slots into a fully aluminium body. With its robust design, this cutter can cope with high cutting frequencies over an extended period of time and has a cutting depth of about 21 mm.
  • Robust handle
  • Double side blade
  • Can be used by left and right-handed users
  • With spring loaded blade retraction, designed to respond to any uncontrolled move if and when the blade slider is not in forward position
Simple blade change by pushing the plastic insert out of the handle and either rotate or replace the blade.
Certificazioni: TÜV tested. Blades are food contact approved.