UV transilluminator bulbs for myECL™ imager
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PIER62255EA 71.25 GBP
PIER62255 PIER62258
UV transilluminator bulbs for myECL™ imager
Photodocumentation Systems
These lamp bulbs with ultraviolet emission are used to replace the one of six pre-installed 306 nm UV bulbs in the myECL™ imager. All six lamps must be the same bulb type (365 or 306 nm).
Order Now
Learn more