Stampa…
Trichrome stain kit (Masson) per istologia, Sigma-Aldrich®
Fornitore: SIGMA-ALDRICH MICROSCOPY
Pericolo
M-Clarity™ quality level = MQ600
HT15-1KTEA 391 EUR
HT15-1KT
Trichrome stain kit (Masson) per istologia, Sigma-Aldrich®
Trichrome stain
This kit may be useful in study of diseases of connective tissue and muscle characterised by fibrotic and dystrophic changes. For in vitro diagnostic use.
Principle: Collagen and muscle fiber stain differentially when treated sequentially with Biebrich Scarlet-Acid Fuchsin, PTA/PMA and Aniline Blue. Cytoplasm and muscle fibers stain red whereas collagen displays blue coloration.
The kit contains 1× Biebrich Scarlet-acid Fuchsin solution (250 ml), 1× phosphomolybdic acid solution (250 ml), 1× phosphotungstic acid solution (250 ml) and 1× aniline blue solution (250 ml).
UN: 3316
ADR: 9,II
IVDLogo