Stampa…
Mobile tablet camera, Moticam® BTX8
Fornitore: Motic
Moticam®
MOTI1100600101051EA 0 EUR
MOTI1100600101051
Mobile tablet camera, Moticam® BTX8
Camere
The Moticam BTX8 combines a space-saving camera hardware with Wi-Fi technology.
  • Pre-installed MotiConnect application to capture, edit and make measurements
  • WiFi signal allows you to connect 5 devices at the same time additionally to the tablet included
  • Can be connected to Ethernet via port RJ 45
Informazioni per l'ordine: Includes a micro SD card, HDMI cable, USB 2.0 cable, Image Plus 3.0 software, 4-dot caibration slide and 100 - 240 V charger.