jetOPTIMUS® DNA Transfection Reagents
Supplier: SARTORIUS
New Product
Certificates
About this item
jetOPTIMUS® is a 'best in class' DNA transfection reagent, based on a novel cationic nanotechnology allowing high efficiency with low volume of reagent.
- Highly efficient: Reach maximum gene expression in hard to transfect cells
- Cost-effective: Use minimum reagent volume and DNA quantity
- Biologically relevant: Keep excellent cell viability and morphology
- Time-saving: Transfect with an optimised, ready to use protocol
jetOPTIMUS® transfection reagent improves cellular uptake and endosomal escape of DNA in cells (even in hard to transfect cells) resulting in higher transfection efficiency. In order to work in relevant physiological conditions, transfection with jetOPTIMUS® requires a minimum DNA quantity and reagent volume to keep excellent cell viability and morphology. Free of charge samples are available upon request. Please contact your local Avantor sales office for further details.
Certifications: Polyplus-transfection® has been an ISO 9001 accredited company since 2002. Since then this certification has been successfully renewed every 3 years.
Specifications
- Cell specific:Mammalian
- CRISPR/Cas 9:Yes
- DNA:Yes
- Storage temperature:5 °C ± 3 °C, stable for at least one year when stored appropriately
Specifications
Related Information
Certificates
Search by Lot Number
Product Information
- Catalog #Supplier #PkAvailabilityPrice
- jetOPTIMUS® DNA transfection reagent (0,1 ml), with 10 ml bufferjetOPTIMUS® DNA transfection reagent (0,1 ml), with 10 ml buffer101000051.1010000510,1 mlLog in for availability1 * 0,1 mL93,00 €Specifications:New ProductNew Product
- jetOPTIMUS® DNA transfection reagent (0,75 ml), with 2×60 ml bufferjetOPTIMUS® DNA transfection reagent (0,75 ml), with 2×60 ml buffer101000025.1010000250,75 mlLog in for availability1 * 0,75 mL630,00 €Specifications:New ProductNew Product
- jetOPTIMUS® DNA transfection reagent (1,5 ml), with 4×60 ml bufferjetOPTIMUS® DNA transfection reagent (1,5 ml), with 4×60 ml buffer101000006.1010000061,5 mlLog in for availability1 * 1,5 mL1 070,00 €Specifications:New ProductNew Product
- jetOPTIMUS® bufferSpecifications:New ProductNew Product