Thermal transfer printer labels, type B-425, WorkHorse™
Supplier: Brady
Certificates
About this item
Printable, polypropylene labels for thermal transfer printers.
- Die-cut labels are supplied on a roll
- Contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates
- Supplied with the ribbon seperate, not in the same enclosure as the tape/sleeves
This material is UL and CSA approved with its respective ribbon. See the recommended ribbon series for the appropriate ribbon to use.
Product Information
- Catalog #Supplier #TypeModelMaterialAdhesivePrinting methodAvailabilityPrice
- Printer labelsPrinter labelsBRDY031326031326B-425THT-55-425-1PPAcrylic1 * 1 Roll177,64 €
Specifications
Environmentally Preferable - Printer labelsPrinter labelsBRDY031327031327B-425THT-76-425-1PPPermanent acrylic1 * 1 Roll260,88 €
Specifications
Environmentally Preferable - Printer labelsPrinter labelsBRDY031328031328B-425THT-19-425-1PPAcrylic1 * 1 Roll137,52 €
Specifications
Environmentally Preferable - Printer labelsPrinter labelsBRDY031321031321B-425THT-7-425-3PPAcrylic1 * 1 Roll237,46 €
Specifications
Environmentally Preferable - Printer labelsPrinter labelsBRDY031322031322B-425THT-17-425-3PPPermanent acrylic1 * 1 Roll137,52 €
Specifications
Environmentally Preferable - Printer labelsPrinter labelsBRDY030196030196B-425THT-19-430-1PPPermanent acrylic1 * 1 Roll139,62 €
Specifications
Environmentally Preferable - Printer labelsPrinter labelsBRDY031320031320B-425THT-5-425-10PPAcrylic1 * 1 Roll149,48 €