Industrial overalls, AlphaTec® 1800 Ts PLUS, model 111
: Ansell
- Catalog No:
- 113-9986
- 113-9981
- 113-9982
- 113-9980
- 113-9983
- 113-9987
- 113-9984
- 113-9985
- Model:
- 111
- 111
- 111
- 111
- 111
- 111
- 111
- 111
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Anti-static:
- +
- +
- +
- +
- +
- +
- +
- +
- Seam type:
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Garment application:
- Chemical protection, General Purpose, Industrial
- Chemical protection, General Purpose, Industrial
- Chemical protection, General Purpose, Industrial
- Chemical protection, General Purpose, Industrial
- Chemical protection, General Purpose, Industrial
- Chemical protection, General Purpose, Industrial
- Chemical protection, General Purpose, Industrial
- Chemical protection, General Purpose, Industrial
- Weight:
- 53 g/m²
- 53 g/m²
- 53 g/m²
- 53 g/m²
- 53 g/m²
- 53 g/m²
- 53 g/m²
- 53 g/m²
- ColourClothing SizePk
- : WH18T-00111-02: ANSEWH18T-00111-02
Specifications
Model111Environmentally PreferableClosure typeZip closureAnti-static+GenderSeam typeBoundGarment applicationChemical protection, General Purpose, IndustrialWeight53 g/m² - : WH18T-00111-03: ANSEWH18T-00111-03
Specifications
Model111Environmentally PreferableClosure typeZip closureAnti-static+GenderSeam typeBoundGarment applicationChemical protection, General Purpose, IndustrialWeight53 g/m² - : WH18T-00111-04: ANSEWH18T-00111-04
Specifications
Model111Environmentally PreferableClosure typeZip closureAnti-static+GenderSeam typeBoundGarment applicationChemical protection, General Purpose, IndustrialWeight53 g/m² - : WH18T-00111-05: ANSEWH18T-00111-05
Specifications
Model111Environmentally PreferableClosure typeZip closureAnti-static+GenderSeam typeBoundGarment applicationChemical protection, General Purpose, IndustrialWeight53 g/m² - : WH18T-00111-06: ANSEWH18T-00111-06
Specifications
Model111Environmentally PreferableClosure typeZip closureAnti-static+GenderSeam typeBoundGarment applicationChemical protection, General Purpose, IndustrialWeight53 g/m² - : WH18T-00111-07: ANSEWH18T-00111-07
Specifications
Model111Environmentally PreferableClosure typeZip closureAnti-static+GenderSeam typeBoundGarment applicationChemical protection, General Purpose, IndustrialWeight53 g/m² - : WH18T-00111-08: ANSEWH18T-00111-08
Specifications
Model111Environmentally PreferableClosure typeZip closureAnti-static+GenderSeam typeBoundGarment applicationChemical protection, General Purpose, IndustrialWeight53 g/m² - : WH18T-00111-09: ANSEWH18T-00111-09
Specifications
Model111Environmentally PreferableClosure typeZip closureAnti-static+GenderSeam typeBoundGarment applicationChemical protection, General Purpose, IndustrialWeight53 g/m²