Stampa…
Glucose, TestStrip™
Fornitore: BioVision
BIOVM1093-200EA 175 EUR
BIOVM1093-200 BIOVM1093-100
Glucose, TestStrip™
TestStrip™ glucose strips are semi-quantitative strips that measures glucose concentration.
These strips are sensitive enough to detect the glucose from the breakdown of starch and are accurate.
Attenzione: Should not be used to diagnose medical conditions such as diabetes.