Zinc chloride, anhydrous 99.99% (trace metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7646-85-7
:
Formula:ZnCl₂
MDL Number:MFCD00011295
:136.3 g/mol
Merck Index:13,10185
Boiling Pt:732 °C (760 mmHg)
Melting Pt:293 °C
Density:2,91 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-592-0
UN:2331
ADR:8,III
:GHS07
:GHS05
:GHS09
Specification Test Results
- Appearance:White to off-white Crystalline powder or crystals
- Total trace metal impurities:≤0.01 %
Specification Test Results