Benzyl benzoate
EDQMB0670000
: EDQM
:
- Pk:1
- Seq:0001
:
:212.25 g/mol
:13,01129
:323 °C (1013 hPa)
:17…20 °C
:1.118 g/cm³ (20 °C)
:158 °C (closed cup)
:Ambient
:204-402-9
:3082
:9,III
:GHS07
:GHS09
:120-51-4
:C₁₄H₁₂O₂
:MFCD00003075