Italy
Order Entry
Benzyl benzoate
  EDQMB0670000
 :  EDQM
 :  
Benzyl benzoate
  EDQMB0670000
 :  EDQM
 :  
245,00 €
  1 * 1 pezzi
This product has been blocked by your organization. Please contact your purchasing department for more information
  • Pk:
    1
  • Seq:
    0001
 :
 :212.25 g/mol
 :13,01129
 :323 °C (1013 hPa)
 :17…20 °C
 :1.118 g/cm³ (20 °C)
 :158 °C (closed cup)
 :Ambient
 :204-402-9
 :3082
 :9,III
 :GHS07
 :GHS09
 :120-51-4
 :C₁₄H₁₂O₂
 :MFCD00003075