
Maleic acid
Catalog # EDQMM0100000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:116.07 g/mol
:13,05726
:256 °C (1013 hPa)
:130…131 °C
:1.59 g/cm³ (20 °C)
:127 °C (closed cup)
:Ambient
:203-742-5
:3261
:8,III
:GHS07
:110-16-7
:C₄H₄O₄
:MFCD00063177