Italy
Order Entry
2-Mercaptoethanol, Millipore®
2-Mercaptoethanol, Millipore®
  444203-250
 :  
undefined
2-Mercaptoethanol, Millipore®
  444203-250
 :  
 : 444203-250
 : CALB444203-250
Price
166,00 €
Log in for availability
Unit of Measure 1 * 250 mL
  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590
 :
 :13,05894
 :157 °C (1013 hPa)
 :–100 °C
 :1.12 g/cm³ (20 °C)
 :68 °C
 :Ambient
 :200-464-6
 :2966
 :6.1,II
 :78.14 g/mol
 :GHS06
 :GHS05
 :GHS09
 :60-24-2
 :HSCH₂CH₂OH
 :MFCD00004890